Allyl isothiocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

377430-100G 377430-500G 377430-5G
SIAL377430-100GEA 85.5 GBP
SIAL377430-100G SIAL377430-500G SIAL377430-5G
Allyl isothiocyanate, Sigma-Aldrich®
Allyl isothiocyanate
Formula: C₄H₅NS
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –80 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004822
CAS Number: 57-06-7
EINECS: 200-309-2
UN: 1545
ADR: 6.1,II
Merck Index: 13,00292

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR