Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

372846-100G 372846-500G
SIAL372846-100GEA 84.5 GBP
SIAL372846-100G SIAL372846-500G
Poly(2-ethyl-2-oxazoline), M.W. ~50,000, Sigma-Aldrich®
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Formula: (C₅H₉NO)x
CAS Number: 25805-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR