Tetraethyl methylenebisphosphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)

M-Clarity™ quality level = MQ200

359181-25ML 359181-5ML
SIAL359181-25MLEA 379 GBP
SIAL359181-25ML SIAL359181-5ML
Tetraethyl methylenebisphosphonate, Sigma-Aldrich®
Tetraethyl methylenebisphosphonate
Formula: C₉H₂₂O₆P₂
MW: 288.22 g/mol
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR