Rhodanine-N-acetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-(4-Oxo-2-thioxothiazolidin-3-yl)acetic acid, Rhodanine-3-acetic acid

M-Clarity™ quality level = MQ100

347558-5G
SIAL347558-5GEA 52.2 GBP
SIAL347558-5G
Rhodanine-N-acetic acid, Sigma-Aldrich®
Rhodanine-N-acetic acid
Formula: C₅H₅NO₃S₂
MW: 191.23 g/mol
MDL Number: MFCD00005491
CAS Number: 5718-83-2
EINECS: 227-220-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR