Barium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

342920-10G 342920-250G 342920-50G
SIAL342920-10GEA 59.3 GBP
SIAL342920-10G SIAL342920-250G SIAL342920-50G
Barium chloride, Sigma-Aldrich®
Barium chloride
Formula: BaCl₂
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: ∼963 °C
Density: 2.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR