1-Bromo-3,4,5-trifluorobenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

330841-5G
SIAL330841-5GEA 41.8 GBP
SIAL330841-5G
1-Bromo-3,4,5-trifluorobenzene, Sigma-Aldrich®
1-Bromo-3,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070738
CAS Number: 138526-69-9
EINECS: 418-480-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR