Manganese(II) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

328146-25G 328146-2KG 328146-500G
SIAL328146-25GEA 39.9 GBP
SIAL328146-25G SIAL328146-2KG SIAL328146-500G
Manganese(II) chloride, Sigma-Aldrich®
Manganese (II) chloride
Formula: MnCl₂
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 650 °C
Density: 2.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05768

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR