Lithium aluminum hydride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

M-Clarity™ quality level = MQ200

323403-100G 323403-25G
SIAL323403-100GEA 223 GBP
SIAL323403-100G SIAL323403-25G
Lithium aluminum hydride, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
Melting Pt: 125 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 13,00348

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR