DNA (Deoxyribonucleic acid), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

31149-10G-F 31149-50G-F
SIAL31149-10G-FEA 66.9 GBP
SIAL31149-10G-F SIAL31149-50G-F
DNA (Deoxyribonucleic acid), Sigma-Aldrich®
DNA (Deoxyribonucleic acid)
MDL Number: MFCD00130921
CAS Number: 9007-49-2
EINECS: 309-566-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR