Zirconium tetrafluoride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Zirconium (IV) fluoride

M-Clarity™ quality level = MQ100

311464-100G 311464-25G
SIAL311464-100GEA 350 GBP
SIAL311464-100G SIAL311464-25G
Zirconium tetrafluoride, Sigma-Aldrich®
Zirconium tetrafluoride
Formula: F₄Zr
Density: 4.43 g/cm³ (20 °C)
MDL Number: MFCD00011307
CAS Number: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8,II
Merck Index: 12,10308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR