Bismuth chloride oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide

M-Clarity™ quality level = MQ100

307610-250G 307610-50G
SIAL307610-250GEA 332 GBP
SIAL307610-250G SIAL307610-50G
Bismuth chloride oxide, Sigma-Aldrich®
Bismuth chloride oxide
Formula: BiClO
Density: 7.72 g/cm³ (20 °C)
MDL Number: MFCD00010895
CAS Number: 7787-59-9
EINECS: 232-122-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR