3,7-Dimethyl-1-octanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,7-Dimethyloctan-1-ol

M-Clarity™ quality level = MQ100

305774-100ML
SIAL305774-100MLEA 80.3 GBP
SIAL305774-100ML
3,7-Dimethyl-1-octanol, Sigma-Aldrich®
3,7-Dimethyl-1-octanol
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 0.849 g/cm³ (0 °C)
MDL Number: MFCD00002936
CAS Number: 106-21-8
EINECS: 203-374-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR