Copper(I) thiocyanate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide

M-Clarity™ quality level = MQ100

298212-250G 298212-50G
SIAL298212-250GEA 299 GBP
SIAL298212-250G SIAL298212-50G
Copper(I) thiocyanate, Sigma-Aldrich®
Copper (I) thiocyanate
Formula: CCuNS
MW: 121.63 g/mol
Density: 2.846 g/cm³ (15 °C)
MDL Number: MFCD00010980
CAS Number: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9 III
Merck Index: 13,02699

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR