Diisopropyl ether, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Isopropyl ether

M-Clarity™ quality level = MQ100

296856-100ML 296856-1L
SIAL296856-100MLEA 78.9 GBP
SIAL296856-100ML SIAL296856-1L
Diisopropyl ether, Sigma-Aldrich®
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
Boiling Pt: 68.5 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.726 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR