Sodium octadecyl sulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt

M-Clarity™ quality level = MQ100

293946-1G
SIAL293946-1GEA 105 GBP
SIAL293946-1G
Sodium octadecyl sulphate, Sigma-Aldrich®
Sodium octadecyl sulphate
Formula: C₁₈H₃₇NaO₄S
MDL Number: MFCD00007469
CAS Number: 1120-04-3
EINECS: 214-295-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR