Trimethylsulphoxonium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Trimethyloxosulphonium chloride

M-Clarity™ quality level = MQ100

293008-5G
SIAL293008-5GEA 60.3 GBP
SIAL293008-5G
Trimethylsulphoxonium chloride, Sigma-Aldrich®
Trimethylsulphoxonium chloride
Formula: C₃H₉ClOS
MDL Number: MFCD00011898
CAS Number: 5034-06-0
EINECS: 225-724-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR