1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1,1-Binaphthyl-2,2-diyl hydrogenphosphate

M-Clarity™ quality level = MQ100

292966-5G
SIAL292966-5GEA 134 GBP
SIAL292966-5G
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula: C₂₀H₁₃O₄P
MW: 348.29 g/mol
MDL Number: MFCD00010045
CAS Number: 35193-63-6
EINECS: 252-425-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR