Magnesium ethanolate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Magnesium ethoxide, Magnesium ethylate

M-Clarity™ quality level = MQ200

291846-100G 291846-500G
SIAL291846-100GEA 96.8 GBP
SIAL291846-100G SIAL291846-500G
Magnesium ethanolate, Sigma-Aldrich®
Magnesium ethanolate
Formula: C₄H₁₀MgO₂
MDL Number: MFCD00009060
CAS Number: 2414-98-4
EINECS: 219-323-5
UN: 3205
ADR: 4.2,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR