Iodoacetic anhydride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

284262-1G 284262-250MG
SIAL284262-1GEA 263 GBP
SIAL284262-1G SIAL284262-250MG
Iodoacetic anhydride, Sigma-Aldrich®
Iodoacetic anhydride
Formula: C₄H₄I₂O₃
MDL Number: MFCD00001080
CAS Number: 54907-61-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR