(R)-(-)-2-Phenylbutyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (R)-(-)-2-Phenylbutanoic acid

M-Clarity™ quality level = MQ200

279870-1G
SIAL279870-1GEA 169 GBP
SIAL279870-1G
(R)-(-)-2-Phenylbutyric acid, Sigma-Aldrich®
(R)-(-)-2-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00063165
CAS Number: 938-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR