Salicylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

M-Clarity™ quality level = MQ200

27301-25KG-R 27301-1KG-R 27301-6X1KG-R
SIAL27301-25KG-REA 1120 GBP
SIAL27301-25KG-R SIAL27301-1KG-R SIAL27301-6X1KG-R
Salicylic acid, Sigma-Aldrich®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1.443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR