L-(-)-tert-Leucine (H-Tle-OH), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH

M-Clarity™ quality level = MQ200

269107-25G 269107-5G
SIAL269107-25GEA 94 GBP
SIAL269107-25G SIAL269107-5G
L-(-)-tert-Leucine (H-Tle-OH), Sigma-Aldrich®
L(-)-tert-Leucine (H-Tle-OH)
Formula: C₆H₁₃NO₂
MDL Number: MFCD00064218
CAS Number: 20859-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR