2,5-Difluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

265187-5G
SIAL265187-5GEA 40.9 GBP
SIAL265187-5G
2,5-Difluorobenzaldehyde, Sigma-Aldrich®
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
Boiling Pt: 65…66 °C (20 mmHg)
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR