1,2-Dibromo-4,5-difluorobenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

264237-10G
SIAL264237-10GEA 156 GBP
SIAL264237-10G
1,2-Dibromo-4,5-difluorobenzene, Sigma-Aldrich®
1,2-Dibromo-4,5-difluorobenzene
Formula: C₆H₂Br₂F₂
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 32.5…33.5 °C
MDL Number: MFCD00009890
CAS Number: 64695-78-9
EINECS: 265-021-1
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR