1-Iodoperfluorododecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Pentacosafluoro-1-iodododecane, Perfluorododecyl iodide

M-Clarity™ quality level = MQ100

257834-1G
SIAL257834-1GEA 77.3 GBP
SIAL257834-1G
1-Iodoperfluorododecane, Sigma-Aldrich®
1-Iodoperfluorododecane
Formula: C₁₂F₂₅I
MW: 745.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001066
CAS Number: 307-60-8
EINECS: 206-205-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR