2-Anisoyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Methoxybenzoyl chloride, 2-Methoxybenzene-1-carbonyl chloride

M-Clarity™ quality level = MQ100

254703-25G 254703-5G
SIAL254703-25GEA 121 GBP
SIAL254703-25G SIAL254703-5G
2-Anisoyl chloride, Sigma-Aldrich®
2-Anisoyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 137 °C (1013 hPa)
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00000664
CAS Number: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR