Sulphanilic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid

M-Clarity™ quality level = MQ200

251917-100G 251917-25G 251917-500G
SIAL251917-100GEA 72 GBP
SIAL251917-100G SIAL251917-25G SIAL251917-500G
Sulphanilic acid, Sigma-Aldrich®
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 279 °C
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR