2-Pyridylacetonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Pyridineacetonitrile

246565-100G 246565-25G 246565-5G
SIAL246565-100GEA 356 GBP
SIAL246565-100G SIAL246565-25G SIAL246565-5G
2-Pyridylacetonitrile, Sigma-Aldrich®
2-Pyridylacetonitrile
Formula: C₇H₆N₂
MDL Number: MFCD00006346
CAS Number: 2739-97-1
EINECS: 220-364-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR