Rubidium hydroxide 50% in aqueous solution, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

243892-25G
SIAL243892-25GEA 345 GBP
SIAL243892-25G
Rubidium hydroxide 50% in aqueous solution, Sigma-Aldrich®
Rubidium hydroxide
Formula: RbOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 300…302 °C
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00011194
CAS Number: 1310-82-3
EINECS: 215-186-0
UN: 2677
ADR: 8,II
Merck Index: 12,08442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR