2-Chlorophenethylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

243736-10G
SIAL243736-10GEA 162 GBP
SIAL243736-10G
2-Chlorophenethylamine, Sigma-Aldrich®
2-Chlorophenethylamine
Formula: C₈H₁₀ClN
MDL Number: MFCD00008185
CAS Number: 13078-80-3
EINECS: 235-982-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR