Allyl chloroformate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

242306-5G 242306-100G
SIAL242306-5GEA 51.5 GBP
SIAL242306-5G SIAL242306-100G
Allyl chloroformate, Sigma-Aldrich®
Allyl chloroformate
Formula: C₄H₅ClO₂
MDL Number: MFCD00000648
CAS Number: 2937-50-0
EINECS: 220-916-6
UN: 1722
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR