Silver(I) sulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Disilver sulphide

M-Clarity™ quality level = MQ100

241474-25G 241474-5G
SIAL241474-25GEA 469 GBP
SIAL241474-25G SIAL241474-5G
Silver(I) sulphide, Sigma-Aldrich®
Silver (I) sulphide
Formula: Ag₂S
Melting Pt: 842 °C
Density: 7.23 g/cm³ (25 °C)
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
Merck Index: 12,08677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR