1,1,1,5,5,5-Hexafluoroacetylacetone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA

M-Clarity™ quality level = MQ100

238309-25G 238309-5G
SIAL238309-25GEA 186 GBP
SIAL238309-25G SIAL238309-5G
1,1,1,5,5,5-Hexafluoroacetylacetone, Sigma-Aldrich®
1,1,1,5,5,5-Hexafluoroacetylacetone
Formula: C₅H₂F₆O₂
Boiling Pt: 70 °C (1013 hPa)
Density: 1.47 g/cm³ (20 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000426
CAS Number: 1522-22-1
EINECS: 216-191-0
UN: 2920
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR