p-Toluidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine

M-Clarity™ quality level = MQ200

236314-25G 236314-100G
SIAL236314-25GEA 46.5 GBP
SIAL236314-25G SIAL236314-100G
p-Toluidine, Sigma-Aldrich®
p-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 201
Melting Pt: 43
Density: 1.046 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00007906
CAS Number: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1 II
Merck Index: 12,09674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR