Sodium ethoxide 21% in ethanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium ethylate, Sodium ethanolate

230553-100ML 230553-500ML
SIAL230553-100MLEA 24.6 GBP
SIAL230553-100ML SIAL230553-500ML
Sodium ethoxide 21% in ethanol, Sigma-Aldrich®
Sodium ethoxide
Formula: C₂H₅NaO
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3274
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR