Aluminium isopropoxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

M-Clarity™ quality level = MQ200

229407-10G 229407-250G 229407-50G
SIAL229407-10GEA 97.2 GBP
SIAL229407-10G SIAL229407-250G SIAL229407-50G
Aluminium isopropoxide, Sigma-Aldrich®
Aluminium isopropoxide
Formula: C₉H₂₁AlO₃
Boiling Pt: 135 °C (13 hPa)
Melting Pt: 119 °C
Density: 0.9771 g/cm³ (20 °C)
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Merck Index: 13,00346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR