2-(Hydroxymethyl)-9,10-anthraquinone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

226521-1G
SIAL226521-1GEA 50 GBP
SIAL226521-1G
2-(Hydroxymethyl)-9,10-anthraquinone, Sigma-Aldrich®
2-(Hydroxymethyl)-9,10-anthraquinone
Formula: C₁₅H₁₀O₃
MDL Number: MFCD00001236
CAS Number: 17241-59-7
EINECS: 241-274-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR