Tris(triphenylphosphine)ruthenium(II) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

223662-1G 223662-25G 223662-5G
SIAL223662-1GEA 72.8 GBP
SIAL223662-1G SIAL223662-25G SIAL223662-5G
Tris(triphenylphosphine)ruthenium(II) chloride, Sigma-Aldrich®
Tris(triphenylphosphine)ruthenium (II) chloride
Formula: C₅₄H₄₅Cl₂P₃Ru
MDL Number: MFCD00013077
CAS Number: 15529-49-4
EINECS: 239-569-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR