Lithium tetrahydroborate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Lithium tetrahydridoborate, Lithium borohydride

M-Clarity™ quality level = MQ100

222356-10G 222356-1G 222356-50G
SIAL222356-10GEA 240 GBP
SIAL222356-10G SIAL222356-1G SIAL222356-50G
Lithium tetrahydroborate, Sigma-Aldrich®
Lithium tetrahydroborate
Formula: H₄BLi
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: 274…277 °C
Density: 0.666 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011088
CAS Number: 16949-15-8
EINECS: 241-021-7
UN: 1413
ADR: 4.3,I
Merck Index: 13,05547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR