1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-Acetoxy-1,3-butadiene

M-Clarity™ quality level = MQ100

220868-10G
SIAL220868-10GEA 218 GBP
SIAL220868-10G
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres), Sigma-Aldrich®
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres)
Formula: C₆H₈O₂
MDL Number: MFCD00008714
CAS Number: 1515-76-0
EINECS: 216-159-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR