(S)-(-)-Limonene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene

MQ100

218367-250G 218367-50G
SIAL218367-250GEA 105 GBP
SIAL218367-250G SIAL218367-50G
(S)-(-)-Limonene, Sigma-Aldrich®
(S)-(-)-Limonene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175
Melting Pt: –74 °C
Density: 0.84 g/cm³ (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00001558
CAS Number: 5989-54-8
EINECS: 227-815-6
UN: 2052
ADR: 3 III
Merck Index: 13,05515

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR