Iron(III) bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

217883-10G 217883-50G
SIAL217883-10GEA 175 GBP
SIAL217883-10G SIAL217883-50G
Iron(III) bromide, Sigma-Aldrich®
Iron (III) bromide
Formula: Br₃Fe
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
UN: 3260
ADR: 8,III
Merck Index: 12,04060

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR