2-Bromo-2-butene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

215562-100G 215562-25G
SIAL215562-100GEA 289 GBP
SIAL215562-100G SIAL215562-25G
2-Bromo-2-butene, Sigma-Aldrich®
2-Bromo-2-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 82
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000141
CAS Number: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR