Copper(I) iodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Cuprous iodide

M-Clarity™ quality level = MQ200

215554-100G 215554-25G 215554-5G
SIAL215554-100GEA 730 GBP
SIAL215554-100G SIAL215554-25G SIAL215554-5G
Copper(I) iodide, Sigma-Aldrich®
Copper (I) iodide
Formula: CuI
Boiling Pt: 1290 °C (1013 hPa)
Melting Pt: 602 °C
Density: 5.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR