Diisobutylaluminium hydride 1 mol/l in dichloromethane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

M-Clarity™ quality level = MQ200

214973-100ML 214973-1L 214973-800ML
SIAL214973-100MLEA 87.5 GBP
SIAL214973-100ML SIAL214973-1L SIAL214973-800ML
Diisobutylaluminium hydride 1 mol/l in dichloromethane, Sigma-Aldrich®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR