2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

210145-100G 210145-10G 210145-25G
SIAL210145-100GEA 102 GBP
SIAL210145-100G SIAL210145-10G SIAL210145-25G
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid), Sigma-Aldrich®
2,2-Dimethyl-1,3-dioxane-4,6-dione (Isopropylidene malonate, Meldrum's acid)
Formula: C₆H₈O₄
MDL Number: MFCD00006638
CAS Number: 2033-24-1
EINECS: 217-992-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR