Dipotassium hexachloroosmate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium hexachloroosmate(IV)

M-Clarity™ quality level = MQ100

209090-1G
SIAL209090-1GEA 175 GBP
SIAL209090-1G
Dipotassium hexachloroosmate, Sigma-Aldrich®
Dipotassium hexachloroosmate
Formula: Cl₆K₂Os
MW: 481.14 g/mol
Melting Pt: 600 °C
Density: 3.42 g/cm³ (20 °C)
MDL Number: MFCD00011371
CAS Number: 16871-60-6
EINECS: 240-893-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR