Ruthenium(III) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ruthenium trichloride

M-Clarity™ quality level = MQ200

208523-10G 208523-2G 208523-50G
SIAL208523-10GEA 662 GBP
SIAL208523-10G SIAL208523-2G SIAL208523-50G
Ruthenium(III) chloride, Sigma-Aldrich®
Ruthenium (III) chloride
Formula: Cl₃Ru
Melting Pt: 500 °C
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00011208
CAS Number: 10049-08-8
EINECS: 233-167-5
UN: 3260
ADR: 8,III
Merck Index: 12,08455

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR