Zirconyl chloride hydrate ≥99.99% (trace metals basis), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Zirconium(IV) dichloride oxide hydrate, Zirconium dichloride oxide hydrate

M-Clarity™ quality level = MQ200

205028-10G 205028-50G
SIAL205028-10GEA 162 GBP
SIAL205028-10G SIAL205028-50G
Zirconyl chloride hydrate ≥99.99% (trace metals basis), Sigma-Aldrich®
Zirconyl chloride hydrate
Formula: Cl₂OZr·H₂O
MDL Number: MFCD00149897
CAS Number: 15461-27-5
EINECS: 231-717-9
UN: 3260
ADR: 8,II
Merck Index: 14,10184

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR