Tellurium dioxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

204552-10G 204552-50G
SIAL204552-10GEA 136 GBP
SIAL204552-10G SIAL204552-50G
Tellurium dioxide, Sigma-Aldrich®
Tellurium dioxide
Formula: O₂Te
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 773 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR